ENA638588090 Enamine

{1-[(oxolan-3-yl)methyl]-1H-1,2,3-triazol-4-yl}methanol

  • MDL number MFCD16747724
  • Cas Number : 1249673-44-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-332707

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-332707
Availability details for
Quantity
  • Availability
  • Add to Cart