ENA638588094 Enamine

2-amino-1-(pyridin-2-yl)propane-1,3-diol dihydrochloride

  • MDL number MFCD30497927
  • Cas Number : 2060024-55-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-332714

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-332714
Availability details for
Quantity
  • Availability
  • Add to Cart