ENA638588128 Enamine

1-chloro-2,4,4-trimethylpentane

  • MDL number MFCD19232362
  • Cas Number : 2371-08-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-332769

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-332769
Availability details for
Quantity
  • Availability
  • Add to Cart