ENA638588171 Enamine

methoxy[(5-methylfuran-2-yl)methyl]amine

  • MDL number MFCD24288874
  • Cas Number : 1554866-90-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-332866

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-332866
Availability details for
Quantity
  • Availability
  • Add to Cart