ENA638588174 Enamine

ethyl 2-(aminomethyl)-2-ethylbutanoate

  • MDL number MFCD16809970
  • Cas Number : 90726-82-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-332872

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-332872
Availability details for
Quantity
  • Availability
  • Add to Cart