ENA638588175 Enamine

(2E)-3-(2,4,6-trimethylphenyl)prop-2-en-1-ol

  • MDL number MFCD21852737
  • Cas Number : 287929-73-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-321990

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-321990
Availability details for
Quantity
  • Availability
  • Add to Cart