ENA638588178 Enamine

(2E)-3-(2,4,5-trimethylphenyl)prop-2-enoic acid

  • MDL number MFCD00075865
  • Cas Number : 205748-06-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-321992

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-321992
Availability details for
Quantity
  • Availability
  • Add to Cart