ENA638588201 Enamine

6-amino-2,4-dimethyl-2,3,4,5-tetrahydro-1,2,4-triazine-3,5-dione

  • MDL number MFCD12103341
  • Cas Number : 1154932-94-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-332927

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-332927
Availability details for
Quantity
  • Availability
  • Add to Cart