ENA638588767 Enamine

3-(3-fluorophenyl)-5-[3-(trifluoromethyl)pyrrolidin-3-yl]-1,2,4-oxadiazole hydrochloride

  • MDL number MFCD30497973
  • Cas Number : 2060041-37-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-332944

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-332944
Availability details for
Quantity
  • Availability
  • Add to Cart