ENA638588772 Enamine

3-(aminomethyl)-3-(trifluoromethyl)pentane hydrochloride

  • MDL number MFCD30497982
  • Cas Number : 2059955-96-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-333034

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-333034
Availability details for
Quantity
  • Availability
  • Add to Cart