ENA638588783 Enamine

azetidin-1-amine dihydrochloride

  • MDL number MFCD30497996
  • Cas Number : 2060060-67-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. -10
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-333128

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-333128
Availability details for
Quantity
  • Availability
  • Add to Cart