ENA638589343 Enamine

1-cyclopropyl-2-(1H-pyrazol-1-yl)ethan-1-one

  • MDL number MFCD19605416
  • Cas Number : 1340400-86-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-333010

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-333010
Availability details for
Quantity
  • Availability
  • Add to Cart