ENA638589344 Enamine

N-[(azetidin-3-yl)methyl]-2,2,2-trifluoroacetamide hydrochloride

  • MDL number MFCD24562003
  • Cas Number : 124668-52-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-333014

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-333014
Availability details for
Quantity
  • Availability
  • Add to Cart