ENA638589365 Enamine

4-methoxy-1,8,8-trimethylbicyclo[3.2.1]oct-3-en-2-one

  • MDL number MFCD01111309
  • Cas Number : 23747-17-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-333458

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-333458
Availability details for
Quantity
  • Availability
  • Add to Cart