ENA638589374 Enamine

tert-butyl N-(3,4-dihydroxybutan-2-yl)carbamate

  • MDL number MFCD24541218
  • Cas Number : 1824519-24-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-333027

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-333027
Availability details for
Quantity
  • Availability
  • Add to Cart