ENA638589395 Enamine

4-ethyl-N,N-dimethylpiperidin-4-amine

  • MDL number MFCD26937960
  • Cas Number : 1823944-23-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-333532

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-333532
Availability details for
Quantity
  • Availability
  • Add to Cart