ENA638589397 Enamine

6-amino-2-(furan-2-yl)pyrimidin-4-ol

  • MDL number MFCD14706103
  • Cas Number : 67945-75-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-333535

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-333535
Availability details for
Quantity
  • Availability
  • Add to Cart