ENA638589406 Enamine

potassium 2-(2-{[(thiophen-2-yl)methyl]amino}-1,3-thiazol-4-yl)acetate

  • MDL number MFCD30498039
  • Cas Number : 2059954-46-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data K+
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-333623

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-333623
Availability details for
Quantity
  • Availability
  • Add to Cart