ENA638589407 Enamine

3-[4-(difluoromethyl)phenyl]propanoic acid

  • MDL number MFCD24225376
  • Cas Number : 1546905-68-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 93-95
Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-333035

Safety Information

signalword Warning
pcodes P261,P264,P270,P272,P280,P301+P312,P302+P352,P305+P351+P338,P330,P333+P313,P337+P313,P363,P501
hcodes Xi,Xn
MSDS https://www.enaminestore.com/catalog/EN300-333035
pictograms GHS07
hstatements H302,H317,H319
Availability details for
Quantity
  • Availability
  • Add to Cart