ENA638589422 Enamine

(1r,4r)-4-(1H-imidazol-2-yl)cyclohexan-1-amine dihydrochloride

  • MDL number MFCD28892742
  • Cas Number : 2059913-83-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-333657

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-333657
Availability details for
Quantity
  • Availability
  • Add to Cart