ENA638589431 Enamine

2-{2-[(4-phenyl-4H-1,2,4-triazol-3-yl)sulfanyl]ethyl}pyridine

  • MDL number MFCD13387705
  • Cas Number : 1210753-31-3
  • Molecular Weight : 282.37

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-333682

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-333682
Availability details for
Quantity
  • Availability
  • Add to Cart