ENA638589439 Enamine

tert-butyl 3-amino-2-methyl-8-azabicyclo[3.2.1]octane-8-carboxylate

  • MDL number MFCD29059019
  • Cas Number : 1895234-36-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-333757

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-333757
Availability details for
Quantity
  • Availability
  • Add to Cart