ENA638589457 Enamine

{2-[4-(benzyloxy)phenyl]ethyl}(methyl)amine hydrochloride

  • MDL number MFCD03695498
  • Cas Number : 24958-41-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-335904

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-335904
Availability details for
Quantity
  • Availability
  • Add to Cart