ENA638589459 Enamine

methyl 5-(pyrrolidin-1-yl)pyridine-3-carboxylate

  • MDL number MFCD22581701
  • Cas Number : 1414866-57-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-335910

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-335910
Availability details for
Quantity
  • Availability
  • Add to Cart