ENA638589465 Enamine

3-ethoxyoxan-4-one

  • MDL number MFCD22049356
  • Cas Number : 624734-18-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-335919

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-335919
Availability details for
Quantity
  • Availability
  • Add to Cart