ENA638589512 Enamine

2-{bicyclo[4.1.0]heptan-3-yl}acetic acid

  • MDL number MFCD28656507
  • Cas Number : 1784988-07-3
  • Molecular Weight : 154.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-335988

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-335988
Availability details for
Quantity
  • Availability
  • Add to Cart