ENA638589518 Enamine

5-bromo-3-methoxy-6-methylpyridin-2-amine

  • MDL number MFCD26407073
  • Cas Number : 947249-16-3
  • Molecular Weight : 217.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 126-130
Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-333155

Safety Information

signalword Warning
pcodes P305+P351+P338, P337+P313,P261,P264,P270,P271,P280,P301+P312,P302+P352,P304+P340,P312,P330,P332+P313,P362,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-333155
pictograms GHS07
hstatements H302,H315,H320,H335
Availability details for
Quantity
  • Availability
  • Add to Cart