ENA638589526 Enamine

2-(1-chloroethyl)-5-(naphthalen-2-yl)-1,3,4-oxadiazole

  • MDL number MFCD12073995
  • Cas Number : 1153824-62-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-333187

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-333187
Availability details for
Quantity
  • Availability
  • Add to Cart