ENA638589550 Enamine

1-(2-bromo-4-chlorophenyl)-1H-1,2,3,4-tetrazole

  • MDL number MFCD24078674
  • Cas Number : 1501463-24-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-333227

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-333227
Availability details for
Quantity
  • Availability
  • Add to Cart