ENA638589559 Enamine

2,5-dimethyloxolan-3-amine hydrochloride

  • MDL number MFCD28411767
  • Cas Number : 1589540-71-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-333249

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-333249
Availability details for
Quantity
  • Availability
  • Add to Cart