ENA638589595 Enamine

1-(3-methyloxolan-3-yl)-1,4-diazepane

  • MDL number MFCD27324883
  • Cas Number : 1597180-38-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-333000

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-333000
Availability details for
Quantity
  • Availability
  • Add to Cart