ENA638589604 Enamine

2-[(3-methylphenyl)methoxy]propanoic acid

  • MDL number MFCD30498011
  • Cas Number : 1603174-94-2
  • Molecular Weight : 194.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-333277

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-333277
Availability details for
Quantity
  • Availability
  • Add to Cart