ENA638589618 Enamine

1-(dimethylamino)butan-2-ol

  • MDL number MFCD06252328
  • Cas Number : 34487-37-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-333296

Safety Information

signalword Danger
pcodes P210,P233,P240,P241,P242,P243,P260,P264,P270,P280,P301+P312,P301+P330+P331,P303+P361+P353,P304+P340,P305+P351+P338,P310,P330,P363,P370+P378,P403+P235,P405,P501
hcodes C,Xn
MSDS https://www.enaminestore.com/catalog/EN300-333296
pictograms GHS02,GHS05,GHS07
hstatements H226,H302,H314
Availability details for
Quantity
  • Availability
  • Add to Cart