ENA638589681 Enamine

4-amino-1,1-difluorobutan-2-ol

  • MDL number MFCD26853454
  • Cas Number : 1538626-71-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-333360

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-333360
Availability details for
Quantity
  • Availability
  • Add to Cart