ENA638589691 Enamine

(3-phenylpiperidin-3-yl)methanol hydrochloride

  • MDL number MFCD30498018
  • Cas Number : 2060037-56-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-333362

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-333362
Availability details for
Quantity
  • Availability
  • Add to Cart