ENA638589757 Enamine

2-(2-phenyl-1H-imidazol-4-yl)acetonitrile

  • MDL number MFCD20676926
  • Cas Number : 52353-66-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-333368

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-333368
Availability details for
Quantity
  • Availability
  • Add to Cart