ENA638594248 Enamine

5-nitro-7-(trifluoromethyl)-1H-indole-2-carboxylic acid

  • MDL number MFCD30500934
  • Cas Number : 2060061-45-0
  • Molecular Weight : 274.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-340363

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-340363
Availability details for
Quantity
  • Availability
  • Add to Cart