ENA638597220 Enamine

2-[(2-methyl-1,3-dioxolan-2-yl)methyl]-1H-imidazole

  • MDL number MFCD30502162
  • Cas Number : 2060040-78-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-341879

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-341879
Availability details for
Quantity
  • Availability
  • Add to Cart