ENA638597222 Enamine

4-amino-3-(4-methylcyclohexyl)butanoic acid hydrochloride

  • MDL number MFCD30502164
  • Cas Number : 2060061-57-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-341901

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-341901
Availability details for
Quantity
  • Availability
  • Add to Cart