ENA638597227 Enamine

7-(difluoromethyl)-1,4-oxazepane hydrochloride

  • MDL number MFCD30502170
  • Cas Number : 2059988-56-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-341946

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-341946
Availability details for
Quantity
  • Availability
  • Add to Cart