ENA638597264 Enamine

[(1-ethyl-1H-1,2,3-triazol-4-yl)methyl](methyl)amine hydrochloride

  • MDL number MFCD30502209
  • Cas Number : 1971106-63-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-342353

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-342353
Availability details for
Quantity
  • Availability
  • Add to Cart