ENA638597274 Enamine

5-bromo-2,6-dichloro-N,N-dimethylpyrimidin-4-amine

  • MDL number MFCD30502222
  • Cas Number : 1656291-41-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-342455

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-342455
Availability details for
Quantity
  • Availability
  • Add to Cart