ENA638597277 Enamine

3-(chloromethyl)-6-fluoroimidazo[1,2-a]pyridine hydrochloride

  • MDL number MFCD30535694
  • Cas Number : 2060032-25-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-342469

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-342469
Availability details for
Quantity
  • Availability
  • Add to Cart