ENA638597284 Enamine

ethyl 3-(aminomethyl)oxane-3-carboxylate hydrochloride

  • MDL number MFCD30535695
  • Cas Number : 2060061-66-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-342527

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-342527
Availability details for
Quantity
  • Availability
  • Add to Cart