ENA638597289 Enamine

2-(2,4-difluorophenyl)-2,2-difluoroethan-1-amine hydrochloride

  • MDL number MFCD30502238
  • Cas Number : 2060062-92-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-342642

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-342642
Availability details for
Quantity
  • Availability
  • Add to Cart