ENA638597296 Enamine

tert-butyl 2-(2,3-dioxoazepan-1-yl)acetate

  • MDL number MFCD30535700
  • Cas Number : 2060006-39-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-342746

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-342746
Availability details for
Quantity
  • Availability
  • Add to Cart