ENA638597596 Enamine

tert-butyl 3-[(dimethylcarbamoyl)amino]-8-azabicyclo[3.2.1]octane-8-carboxylate

  • MDL number MFCD30535703
  • Cas Number : 2060063-02-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-343097

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-343097
Availability details for
Quantity
  • Availability
  • Add to Cart