ENA638597612 Enamine

methyl 4-amino-3-(2-hydroxyethoxy)benzoate hydrochloride

  • MDL number MFCD30535723
  • Cas Number : 2060007-27-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-343270

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-343270
Availability details for
Quantity
  • Availability
  • Add to Cart