ENA638597647 Enamine

1-(2,4-dimethoxyphenyl)piperazine

  • MDL number MFCD00040794
  • Cas Number : 16015-75-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-342304

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-342304
Availability details for
Quantity
  • Availability
  • Add to Cart