ENA638597660 Enamine

3-(cycloheptylamino)butanenitrile

  • MDL number MFCD12812210
  • Cas Number : 1178057-99-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-342357

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-342357
Availability details for
Quantity
  • Availability
  • Add to Cart